Learn More
Crescent Chemical Co Inc SERVA DNA STAIN G
SDP
Catalog No. 50248493
Encompass_Preferred
SERVA DNA Stain G SERVA DNA Stain G is a safer alternative to traditional ethidium bromide stain for detecting nucleic acids in agarose gels. It is at least as sensitive as ethidium bromide and can be used in exactly the same way in agarose gel electrophoresis. SE
Catalog No. 50-248-493 Supplier Crescent Chemical Co Inc Supplier No. SE39803.01
SERVA DNA Stain G SERVA DNA Stain G is a safer alternative to traditional ethidium bromide stain for detecting nucleic acids in agarose gels. It is at least as sensitive as ethidium bromide and can be used in exactly the same way in agarose gel electrophoresis. SE