Learn More
Crescent Chemical Co Inc SERVA DNA STD 100 BP LADDER
SDP
Catalog No. 50248373
Encompass_Preferred
SERVA DNA Standard 100 Bp ladder extended, lyophilized The SERVA DNA Standard 100 bp ladder extended contains 17 fragments ranging from 100 bp to 5000 bp:100, 150, 200, 300, 400, 500 (2x), 600, 700, 800, 900, 1000, 1500, 2000, 2500, 3000, 4000 and 5000 bp.Ideal for the analysis of DNA fragments gener
Catalog No. 50-248-373 Supplier Crescent Chemical Co Inc Supplier No. SE39312.01
SERVA DNA Standard 100 Bp ladder extended, lyophilized The SERVA DNA Standard 100 bp ladder extended contains 17 fragments ranging from 100 bp to 5000 bp:100, 150, 200, 300, 400, 500 (2x), 600, 700, 800, 900, 1000, 1500, 2000, 2500, 3000, 4000 and 5000 bp.Ideal for the analysis of DNA fragments gener