Learn More
Crescent Chemical Co Inc SERVA DNA STD 100 BP LADDER
SDP
Supplier:  Crescent Chemical Co Inc SE39311.01
Encompass_Preferred
SERVA DNA Standard 100 Bp ladder equimolar lyophilized The SERVA DNA Standard 100 bp ladder equimolar contains 11 fragments ranging from 100 bp to 1000 bp:100, 150, 200, 300, 400, 500 (2x), 600, 700, 800, 900 and 1000 bp.Ideal for the analysis of DNA fragments generated from plasmid DNA or
Catalog No. 50-248-372