Learn More
Crescent Chemical Co Inc SERVA TRIS-GLYCINE/SDS BUFFER
SDP
Catalog No. 50248591
Encompass_Preferred
SERVA Tris-Glycine/SDS sample buffer (2x) Sample buffer for SDS PAGE. Supplied as 2 x concentrate. Contains 126 mM Tris/HCl (pH 6.8), 20 % glycerol, 4 % SDS and 0.02 % bromophenol blue. Size - 20ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42527.01
Catalog No. 50-248-591 Supplier Crescent Chemical Co Inc Supplier No. SE42527.01
Add to Cart
Add to Cart
SERVA Tris-Glycine/SDS sample buffer (2x) Sample buffer for SDS PAGE. Supplied as 2 x concentrate. Contains 126 mM Tris/HCl (pH 6.8), 20 % glycerol, 4 % SDS and 0.02 % bromophenol blue. Size - 20ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42527.01