Learn More
Crescent Chemical Co Inc SERVA TRIS-TRICINE/SDS BUFFER
SDP
Catalog No. 50248605
Encompass_Preferred
SERVA Tris-Tricine/SDS electrophoresis buffer (10x) Running buffer for SDS PAGE. Supplied as 10 x concentrate. Contains 1 M Tris, 1 M Tricine and 1 % SDS. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 42552.01
Catalog No. 50-248-605 Supplier Crescent Chemical Co Inc Supplier No. SE42552.01
SERVA Tris-Tricine/SDS electrophoresis buffer (10x) Running buffer for SDS PAGE. Supplied as 10 x concentrate. Contains 1 M Tris, 1 M Tricine and 1 % SDS. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 42552.01