Learn More
Crescent Chemical Co Inc SERVA TRIS-TRICINE/SDS BUFFER
SDP
Supplier:  Crescent Chemical Co Inc SE42560.01
Encompass_Preferred
SERVA Tris-Tricine/SDS electrophoresis buffer (20x) Running buffer for SDS PAGE. Optimized formulation to improve sharpness of bands, especially when used together with SERVAGel Neutral pH 7.4 (cat. no. 43220 and 43221) precast gels. Supplied as 20 x concentrate. Contains 1.2 M Tris (pH 8.5), 0.8
Catalog No. 50-248-616