Learn More
Crescent Chemical Co Inc SERVA VIOLET 17
SDP
Catalog No. 50248125
Encompass_Preferred
SERVA Violet 17 For staining of proteins in PAGE. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35072.02 CAS 4129-84-4
Catalog No. 50-248-125 Supplier Crescent Chemical Co Inc Supplier No. SE35072.02
SERVA Violet 17 For staining of proteins in PAGE. Size - 25G Storage Conditions - +15 °C TO +30 °C Catalog Number - 35072.02 CAS 4129-84-4