Learn More
Crescent Chemical Co Inc SERVALYT™ 2-4
SDP
Catalog No. 50248647
Encompass_Preferred
SERVALYT™ 2-4 40 % (w/v) solution in water SERVALYT™ Carrier Ampholytes (40 % w/v in water). Standard quality (analytical grade) for general use in IEF. Size - 10ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42902.01
Catalog No. 50-248-647 Supplier Crescent Chemical Co Inc Supplier No. SE42902.01
SERVALYT™ 2-4 40 % (w/v) solution in water SERVALYT™ Carrier Ampholytes (40 % w/v in water). Standard quality (analytical grade) for general use in IEF. Size - 10ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42902.01