Learn More
Crescent Chemical Co Inc SERVALYT™ 2-9 SEED-MIX
SDP
Supplier:  Crescent Chemical Co Inc SE42935.03
Encompass_Preferred
SERVALYT™ 2-9 Seed-Mix 40 % (w/v) solution in water SERVALYT™ Carrier Ampholytes (40 % w/v in water). Special quality for seed analysis by IEF. Size - 100ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42935.03
Catalog No. 50-248-689