Learn More
Crescent Chemical Co Inc SERVALYT™ 2-9 SEED-MIX
SDP
Catalog No. 50248688
Encompass_Preferred
SERVALYT™ 2-9 Seed-Mix 40 % (w/v) solution in water SERVALYT™ Carrier Ampholytes (40 % w/v in water). Special quality for seed analysis by IEF. Size - 25ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42935.02
Catalog No. 50-248-688 Supplier Crescent Chemical Co Inc Supplier No. SE42935.02
SERVALYT™ 2-9 Seed-Mix 40 % (w/v) solution in water SERVALYT™ Carrier Ampholytes (40 % w/v in water). Special quality for seed analysis by IEF. Size - 25ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42935.02