Learn More
Crescent Chemical Co Inc SERVALYT™ 3-10 ISO-DALT
SDP
Supplier:  Crescent Chemical Co Inc SE42951.01
Encompass_Preferred
SERVALYT™ 3-10 Iso-Dalt 40 % (w/v) solution in water Iso-Dalt quality; special 2D grade to be used in 2D electrophoresis. Size - 10ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42951.01
Catalog No. 50-248-708