Learn More
Crescent Chemical Co Inc SERVALYT™ 3-10 ISO-DALT
SDP
Catalog No. 50248710
Encompass_Preferred
SERVALYT™ 3-10 Iso-Dalt 40 % (w/v) solution in water Iso-Dalt quality; special 2D grade to be used in 2D electrophoresis. Size - 2ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42951.04
Catalog No. 50-248-710 Supplier Crescent Chemical Co Inc Supplier No. SE42951.04
SERVALYT™ 3-10 Iso-Dalt 40 % (w/v) solution in water Iso-Dalt quality; special 2D grade to be used in 2D electrophoresis. Size - 2ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42951.04