Learn More
Crescent Chemical Co Inc SERVALYT™ 3-10 ISO-DALT
Supplier: Crescent Chemical Co Inc SE42951.02
SERVALYT™ 3-10 Iso-Dalt 40 % (w/v) solution in water Iso-Dalt quality; special 2D grade to be used in 2D electrophoresis. Size - 25ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42951.02