Learn More
Crescent Chemical Co Inc SERVALYT™ 4-9 T
SDP
Supplier:  Crescent Chemical Co Inc SE42910.01
Encompass_Preferred
SERVALYT™ 4-9 T 40 % (w/v) solution in water Technical grade quality for preparative work. Formation of solid particle (quart. ammonium salts) can be found over time of storage at low temperature. This will not affect the separation as precipitate will dissolve up
Catalog No. 50-248-666