Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crescent Chemical Co Inc Servalyt 8-10 | 10mL
SDP

Catalog No. NC1541144
Encompass_Preferred

Servalyt 8-10 | 40 % (w/v) solution in water | 10mL

Catalog No. NC1541144 Supplier Crescent Chemical Co Inc Supplier No. 42911.01
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Servalyt 8-10 | 40 % (w/v) solution in water | 10mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.