Learn More
Crescent Chemical Co Inc SERVAPOR DIALYSIS TUBING
SDP
Catalog No. 50248865
Encompass_Preferred
SERVAPOR dialysis tubing, MWCO 12 000 - 14 000 RC, diameter 29 mm Packed dry , made from regenerated cellulose (RC). With glycerol as protection for embrittlement, which can be easily removed by soaking in water. Contain low level of heavy metal and sulfide impurities. Highly resistant against
Catalog No. 50-248-865 Supplier Crescent Chemical Co Inc Supplier No. SE44146.04
SERVAPOR dialysis tubing, MWCO 12 000 - 14 000 RC, diameter 29 mm Packed dry , made from regenerated cellulose (RC). With glycerol as protection for embrittlement, which can be easily removed by soaking in water. Contain low level of heavy metal and sulfide impurities. Highly resistant against