Learn More
Crescent Chemical Co Inc SERVAPOR DIALYSIS TUBING
SDP
Supplier:  Crescent Chemical Co Inc SE44144.01
Encompass_Preferred
SERVAPOR dialysis tubing, MWCO 12 000 - 14 000 RC, diameter 21 mm Packed dry , made from regenerated cellulose (RC). With glycerol as protection for embrittlement, which can be easily removed by soaking in water. Contain low level of heavy metal and sulfide impurities. Highly resistant against
Catalog No. 50-248-860