Learn More
Hach Company Service Bracket for TU5
Supplier:  Hach Company LZY873
Encompass
Servicing your TU5 series turbidimeter is easy with the service bracket accessory The service bracket can hold the process head with or without the automatic cleaning module the vial removal tool and a manual wiper making manual cleaning and vial replacement a simple task
Catalog No. 50-002-07595