Learn More
Mettler Toledo™ Service for Entry LA Balances
Catalog No. 01258280 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Service Offering:
IPac Balances with Accredited ACC
StarterPac Installation with ACC
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Service Offering
01-258-280 IPac Balances with Accredited ACC
01-258-285 StarterPac Installation with ACC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-280 Supplier Mettler Toledo™ Supplier No. B99484130131
Add to Cart
Add to Cart
Professional installation completed by a trained service technician will guarantee maximum performance and optimal use.
To achieve peak performance and compliance with quality systems and regulatory requirements, take the right steps at the beginning of equipment use.
  • The Accuracy Calibration Certificate (ACC) is the new benchmark in the calibration of weighing equipment.
  • Tests include repeatability, eccentricity and error of idication.
  • A statement of measurement uncertainty is also included.
  • StarterPac includes: Professional installation performed by a trained and authorized service technician, installation confirmation/documentation, immediate deployment and use of your equipment, professionally trained operating personnel
  • IPac includes: Documented standard qualification and initial calibration, compliance with internal and external regulations, immediate deployment for quick, safe release to routine operation, and improved productivity with correct equipment set-up and initial user training
  • EQPac includes: Regulatory conformance qualification documentation, professional qualification including test procedures, specification of weighing ranges and calibration, immediate deployment for quick, safe release to begin routine operation, improved productivity with correct equipment set-up and initial user training, cost savings through the use of a manufacturer developed qualification protocol
  • Extended Warranty: Extends manufacturer's warranty by an additional 12 months
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Entry Analytical and Precision LA Balances
Product Type Balance Service
Service Offering IPac Balances with Accredited ACC