Learn More
BRANDTECH™ VACUUBRAND™ Service Kits for Oil-Free Diaphragm Vacuum Pumps
Catalog No. 13889107 Shop All BRANDTECH Scientific Products
Change view
Click to view available options
For Use With:
MD 4 NT/VARIO, MV 2 NT/VARIO
MD1C, PC3001 Diaphragm Pump
ME 8 NT
ME1, ME1C Diaphragm Pump
ME2C NT Diaphragm Pump
MZ 2 NT, ME 4 NT + VARIO
MZ2D NT VARIO
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
13-889-107 ME1, ME1C Diaphragm Pump
13-688-970 MZ2D NT VARIO
13-889-108 ME2C NT Diaphragm Pump
13-688-969 ME 8 NT
13-688-968 MD 4 NT/VARIO, MV 2 NT/VARIO
13-688-967 MZ 2 NT, ME 4 NT + VARIO
13-889-106 MD1C, PC3001 Diaphragm Pump
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 13-889-107 Supplier BRANDTECH™ VACUUBRAND™ Supplier No. 20696874
Add to Cart
Edge
Add to Cart
Original VACUUBRAND replacement parts
  • Includes appropriate diaphragms, valves, and sealing rings, if appropriate
  • Does not include spanner wrench
TRUSTED_SUSTAINABILITY
Specifications
For Use With ME1, ME1C Diaphragm Pump
Type Service Kit
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov