Description
To achieve peak performance and compliance with quality systems and regulatory requirements, take the right steps at the beginning of equipment use.
- The Accuracy Calibration Certificate (ACC) is the new benchmark in the calibration of weighing equipment.
- Tests include repeatability, eccentricity and error of idication.
- A statement of measurement uncertainty is also included.
- StarterPac includes: Professional installation performed by a trained and authorized service technician, installation confirmation/documentation, immediate deployment and use of your equipment, professionally trained operating personnel
- IPac includes: Documented standard qualification and initial calibration, compliance with internal and external regulations, immediate deployment for quick, safe release to routine operation, and improved productivity with correct equipment set-up and initial user training
- EQPac includes: Regulatory conformance qualification documentation, professional qualification including test procedures, specification of weighing ranges and calibration, immediate deployment for quick, safe release to begin routine operation, improved productivity with correct equipment set-up and initial user training, cost savings through the use of a manufacturer developed qualification protocol
- Extended Warranty: Extends manufacturer's warranty by an additional 12 months
Specifications
Specifications
| For Use With (Equipment) | Standard MR, MA Analytical Balances |
| Product Type | Balance Service |
| Service Offering | IPac Balances with Accredited ACC |