Learn More
Grainger Serving Spoon 9 in L Red PK12
Catalog No. 502752453
Encompass_Preferred
The SP Scienceware Sampler Spoon is a stainless steel spoon designed for sampling powders and granular materials. It has a bowl capacity of 5mL and a total length of 4 inches. The spoon is made from 304 stainless steel for durability and easy cleaning. It can be used for sampling applications in laboratories, industrial facilities, food manufacturing, and more. The sampler spoon allows users to scoop up precise amounts of material for testing or inspection. Its small size is ideal for obtaining representative samples from bags, drums, batches, or other containers. This durable stainless steel spoon provides an economical and effective way to collect material samples.
Catalog No. 50-275-2453 Supplier Grainger Supplier No. 14D264
The SP Scienceware Sampler Spoon is a stainless steel spoon designed for sampling powders and granular materials. It has a bowl capacity of 5mL and a total length of 4 inches. The spoon is made from 304 stainless steel for durability and easy cleaning. It can be used for sampling applications in laboratories, industrial facilities, food manufacturing, and more. The sampler spoon allows users to scoop up precise amounts of material for testing or inspection. Its small size is ideal for obtaining representative samples from bags, drums, batches, or other containers. This durable stainless steel spoon provides an economical and effective way to collect material samples.