Learn More
V&P Scientific Inc Vertical Tumble Stirrer, Double Stack Servo Motor, 5 SLAS Positions, Manual Control
Catalog No. NC2135384
Encompass
STIRRER, Vertical Tumble, NEMA 34 Servo Motor, High Speed and High Torque, 48 MGO NdFeB Magnetic Drive Cylinder, 54cm Long Usable Stirring Deck, Stirs 5 Deep Well Microplates, includes Adjustable Feet, Low Profile Deck made from ABS, with RPM meter on LCD, IPC-5 75 Volt Power Supply, 120/220 Volts, 50/60 Hz, CE Compliant
This item is not returnable. View return policy
Catalog No. NC2135384 Supplier V&P Scientific Inc Supplier No. VP 710E5X
Add to Cart
Add to Cart
Request a Quote
STIRRER, Vertical Tumble, NEMA 34 Servo Motor, High Speed and High Torque, 48 MGO NdFeB Magnetic Drive Cylinder, 54cm Long Usable Stirring Deck, Stirs 5 Deep Well Microplates, includes Adjustable Feet, Low Profile Deck made from ABS, with RPM meter on LCD, IPC-5 75 Volt Power Supply, 120/220 Volts, 50/60 Hz, CE Compliant