Learn More
Honeywell™ Servus™ PRM Safety Knee Boot
Catalog No. 19920150
Change view
Click to view available options
Size:
12
13
14
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-920-150 12
19-920-151 13
19-920-152 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-920-150 Supplier Honeywell™ Supplier No. 75145CBLK120
This item is not returnable. View return policy
Seamless, injection-molded footwear is 100 percent waterproof to ensure feet are dry and comfortable
  • Flexible steel midsoles provide under-foot puncture protection
  • Open pattern outsole design self-cleans liquid and debris with every step, helping to prevent slip and trip hazards
  • Removable FOOT FORM contour cushion insoles provide all-day comfort and support
  • Available in US men sizes 7 to 14
  • Hi Boot (38.1cm/15 in.)
  • Steel toe
Compliance
Meets ASTM F2413-11 M I/75 C/75 and PR.
TRUSTED_SUSTAINABILITY
Specifications
Height (English) 15 in.
Size 12