Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Servus™ PRO™ PVC Knee Boots

Catalog No. 19121552F
Change view
Click to view available options
Size:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-552F 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-121-552F Supplier Honeywell™ Supplier No. 75102GYM100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Honeywell™ Servus™ PRO™ PVC Knee Boots with Triple Density Technology™ that combines three uniquely formulated compounds, injection-molded to form a rugged, high performance boot with outstanding comfort and durability.

  • Perimeter of the outsole is formed from a high-density compound that resists abrasion
  • Midsole and interior of the outsole is softer to offer superior grip and comfort
  • A third compound, formulated for flexibility and fit, forms the upper part of the boot; it provides additional abrasion and chemical resistance
  • Seamless construction is 100% waterproof
  • Foot Form™ replaceable contoured cushion insole
  • Steel shank is longer, larger and positioned for optimal support
  • High, wide kick-off lug
  • Self-cleaning triangular cleats
  • Style 75101 boots also feature Safe-Toe™ steel toe cap
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Meets ASTM™ F2413-11 M I/75 C/75
Color Gray
For Use With (Application) Agriculture, Food Services, Government, Manufacturing, Nuclear, Oil And Gas, Utilities
Height (English) 15 in.
Height (Metric) 38.1 cm
Material PVC
Seam Type Seamless
Size 10
Sole Material TDT™ Dual Compound Yellow And Beige Outsole
Style Knee Boots
Toe Style Plain Toe
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.