Learn More
Honeywell™ Servus™ Puncture-Resistant Midsole
Catalog No. 199201768
Change view
Click to view available options
Size:
8
9
10
11
12
13
14
15
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-920-1768 15
19-920-1761 8
19-920-1762 9
19-920-1763 10
19-920-1764 11
19-920-1765 12
19-920-1767 14
19-920-1766 13
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 19-920-1768 Supplier Honeywell™ Supplier No. 29000BLK150
This item is not returnable. View return policy
Cushion-covered, flexible steel inserts provide puncture protection underfoot
  • Use as an insole or midsole
  • Will fit most over-the-foot boots
TRUSTED_SUSTAINABILITY
Specifications
Material Cushion
Size 15