Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sesame oil, MP Biomedicals™

Catalog No. ICN15662191 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15662191 1 L
ICN15662183 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15662191 Supplier MP Biomedicals, Inc Supplier No. 0215662191
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8008-74-0
Synonym Benne oil, Teel oil

Specifications

Density 0.915-0.925g/mL
Quantity 1 L
Physical Form Oily Liquid
Chemical Name or Material Sesame oil
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.