Learn More
Medchemexpress LLC HY-N0121 5mg Medchemexpress, Sesamin CAS:607-80-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN01215MG
Encompass_Preferred
Medchemexpress, HY-N0121 5mg Sesamin CAS:607-80-7 Sesamin, abundant lignan found in sesame oil, is a potent and selective delta 5 desaturase inhibitor in polyunsaturated fatty acid biosynthesis. Sesamin exerts effective neuroprotection against cerbral ischemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0225