Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sesamin, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014866 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4866 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4866 Supplier MilliporeSigma Supelco Supplier No. 5986710MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity analytical standard of the non-competitive Δ5-desaturase inhibitor Sesamin.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sesamin
CAS 607-80-7
Physical Form Neat
Linear Formula C20H18O6
Technique HPLC
MDL Number MFCD00216134
Synonym Fagarol
Type Analytical Standard
Industry Type Food Testing
Molecular Weight (g/mol) 354.35
Quantity 10 mg
Formula Weight 354.35
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.