Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sesamin, Thermo Scientific Chemicals

Catalog No. AC464892500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464892500 250 mg
AC464890010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464892500 Supplier Thermo Scientific Chemicals Supplier No. 464892500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 607-80-7 | C20H18O6 | 354.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-80-7
Molecular Formula C20H18O6
Molecular Weight (g/mol) 354.36
InChI Key PEYUIKBAABKQKQ-AFHBHXEDSA-N
IUPAC Name 5-[(1S,3aR,4S,6aR)-4-(2H-1,3-benzodioxol-5-yl)-hexahydrofuro[3,4-c]furan-1-yl]-2H-1,3-benzodioxole
SMILES C1OC2=CC=C(C=C2O1)[C@H]1OC[C@H]2[C@@H]1CO[C@@H]2C1=CC=C2OCOC2=C1

Specifications

Melting Point 123.0°C
Quantity 250 mg
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in chloroform
Formula Weight 354.36
Chemical Name or Material Sesamin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.