Learn More
Medchemexpress LLC Sesamol | 533-31-3 | 138.12 | 500 G
SDP
Catalog No. 5000415800
Encompass_Preferred
Sesamol, a constituent of sesame oil, exhibits free radical scavenging activity and shows anti-oxidant and anticancer activities. It can induce apoptosis in human liver hepatocellular carcinoma (HepG2) cells by impairing mitochondrial function and suppressing autophagy.
  • Exhibits free radical scavenging, anti-oxidant, and anticancer activities.
  • Induces apoptosis in human liver hepatocellular carcinoma (HepG2) cells.
  • Impairs mitochondrial function and suppresses autophagy.
  • Decreases anti-apoptotic protein Bcl-2 expression.
  • Improves Fas/FasL protein expression, activating tBid and caspase-8.
  • Inhibits mitophagy and autophagy via the PI3K Class III/Belin-1 pathway.
  • Significantly inhibits tumor volume and down-regulates cell proliferation marker Ki76 in vivo.
  • Increases cell apoptosis marker cleaved-caspase 3.
Catalog No. 50-004-15800 Supplier Medchemexpress LLC Supplier No. HYN1417500G
Sesamol, a constituent of sesame oil, exhibits free radical scavenging activity and shows anti-oxidant and anticancer activities. It can induce apoptosis in human liver hepatocellular carcinoma (HepG2) cells by impairing mitochondrial function and suppressing autophagy.
  • Exhibits free radical scavenging, anti-oxidant, and anticancer activities.
  • Induces apoptosis in human liver hepatocellular carcinoma (HepG2) cells.
  • Impairs mitochondrial function and suppresses autophagy.
  • Decreases anti-apoptotic protein Bcl-2 expression.
  • Improves Fas/FasL protein expression, activating tBid and caspase-8.
  • Inhibits mitophagy and autophagy via the PI3K Class III/Belin-1 pathway.
  • Significantly inhibits tumor volume and down-regulates cell proliferation marker Ki76 in vivo.
  • Increases cell apoptosis marker cleaved-caspase 3.