Learn More
TARGETMOL CHEMICALS INC SESAMOLIN 10MG
SDP
Catalog No. 502576632
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Sesamolin and Sesamin has neuroprotective effect. 2. Sesamolin protects microglia against H2O2-induced cell injury by inhibiting of p38 MAPK and caspase-3 activation and ROS production. 3. Sesamolin and Sesamin can significantly attenuate the excess generation of nitric oxide in lipopolysaccharide-stimulated rat primary microglia cells. purity: 98%
Catalog No. 50-257-6632 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S228310MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Sesamolin and Sesamin has neuroprotective effect. 2. Sesamolin protects microglia against H2O2-induced cell injury by inhibiting of p38 MAPK and caspase-3 activation and ROS production. 3. Sesamolin and Sesamin can significantly attenuate the excess generation of nitric oxide in lipopolysaccharide-stimulated rat primary microglia cells. purity: 98%