Learn More
Medchemexpress LLC Sesamolin | 526-07-8 | 99.8% | 370.35 | C20H18O7 | 10 MG
SDP
Catalog No. 5000296450
Encompass_Preferred
Sesamolin is a lignan isolated from Sesamum indicum with antioxidative and neuroprotective activity. It inhibits lipid peroxidation and suppresses MAPK cascade signaling by preventing phosphorylation of JNK, p38 MAPKs, and caspase-3; oral activity has been reported. The compound is supplied as a solid and as DMSO solutions and is typically provided at high purity for research use.
  • Antioxidative activity
  • Inhibits lipid peroxidation
  • Suppresses JNK and p38 MAPK phosphorylation
  • Reduces caspase-3 activation
  • Reported oral bioactivity
  • High purity (99.78%)
  • Available as solid and DMSO solution, including 10 mg solid
Catalog No. 50-002-96450 Supplier Medchemexpress LLC Supplier No. HYN080910MG
Sesamolin is a lignan isolated from Sesamum indicum with antioxidative and neuroprotective activity. It inhibits lipid peroxidation and suppresses MAPK cascade signaling by preventing phosphorylation of JNK, p38 MAPKs, and caspase-3; oral activity has been reported. The compound is supplied as a solid and as DMSO solutions and is typically provided at high purity for research use.
  • Antioxidative activity
  • Inhibits lipid peroxidation
  • Suppresses JNK and p38 MAPK phosphorylation
  • Reduces caspase-3 activation
  • Reported oral bioactivity
  • High purity (99.78%)
  • Available as solid and DMSO solution, including 10 mg solid