Learn More
Medchemexpress LLC HY-N0809 5mg Medchemexpress, Sesamolin CAS:526-07-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN08095MG
Encompass_Preferred
Medchemexpress, HY-N0809 5mg Sesamolin CAS:526-07-8 Sesaminol, isolated from Justicia orbiculata, has antioxidative activity, Sesaminol inhibits lipid peroxidation and shows neuroprotection effect. Sesaminol potently inhibits MAPK cascades by preventing phosphorylation of JNK, p38 MAPKs, and caspase-3 but not ERK-MAPK expression[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2823