Learn More
Aviva Systems Biology Corp SESN2 Antibody - N-terminal region (ARP60639_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60639_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SESN2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SESN2. Alternate names: DKFZp761M0212, DKFZp761M02121, HI95, SES2, SEST2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig
Catalog No. 50-287-109