Learn More
TARGETMOL CHEMICALS INC SET 2 5MG
SDP
Catalog No. 502580147
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SET 2 is an antagonist of transient receptor potential vanilloid type 2(TRPV2) with an IC50 of 0.46 uM. SET 2 shows selectivity over TRPV1 TRPV3 and TRPV4. purity: 98%
Catalog No. 50-258-0147 Supplier TARGETMOL CHEMICALS INC Supplier No. T370975MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. SET 2 is an antagonist of transient receptor potential vanilloid type 2(TRPV2) with an IC50 of 0.46 uM. SET 2 shows selectivity over TRPV1 TRPV3 and TRPV4. purity: 98%