Learn More
Fritsch Milling And Sizing Inc PULVERISETTE 14 Premium Line Set Sieve Shells 4 mm Trapezoidal Perforation, Stainless Steel
SDP
Catalog No. 501533997
Encompass
Set of sieve shells 4 mm round perforation made of stainless steel for PULVERISETTE 14 premium line.
With a few motions turn your Centrifugal Mill into a Cutting Mill for fast, efficient pre-grinding of fibrous materials and plastics.
Simply insert the cutting rotor with cooling fins with the appropriate accessories such as the labyrinth disk, collecting vessel, holder for sieve shells and a set of sieve shells – the materials are ground by cutting and shearing forces.
The desired final fineness will also be determined here by the selected sieve shells with trapezoidal or round perforation.
Catalog No. 50-153-3997 Supplier Fritsch Milling And Sizing Inc Supplier No. 14.4564.00
Set of sieve shells 4 mm round perforation made of stainless steel for PULVERISETTE 14 premium line.
With a few motions turn your Centrifugal Mill into a Cutting Mill for fast, efficient pre-grinding of fibrous materials and plastics.
Simply insert the cutting rotor with cooling fins with the appropriate accessories such as the labyrinth disk, collecting vessel, holder for sieve shells and a set of sieve shells – the materials are ground by cutting and shearing forces.
The desired final fineness will also be determined here by the selected sieve shells with trapezoidal or round perforation.