Learn More
Med Vet International Vacutainer Blood Collection Set, 23g X .75" Lt Blue, 50 Ea/Box
Catalog No. 502830439
Encompass_Preferred
Vacutainer Safety-Lok Blood Collection and Infusion Set with Luer Adapter, 23G X 0.75" Needle X 12" Tubing. Manually activated safety shield to fully cover needle. BD Safety-Lok™ Blood Collection Sets have been designed to shield healthcare workers from exposed needles. Following venipuncture, a yellow transparent shield is moved over the exposed needle and locked into protective position, creating a barrier between the healthcare professional and the used needle.
Catalog No. 50-283-0439 Supplier Med Vet International Supplier No. BD367283Z
 Restricted Item
Add to Cart
Add to Cart
Vacutainer Safety-Lok Blood Collection and Infusion Set with Luer Adapter, 23G X 0.75" Needle X 12" Tubing. Manually activated safety shield to fully cover needle. BD Safety-Lok™ Blood Collection Sets have been designed to shield healthcare workers from exposed needles. Following venipuncture, a yellow transparent shield is moved over the exposed needle and locked into protective position, creating a barrier between the healthcare professional and the used needle.