Learn More
Electron Microscopy Sciences 30 Slide Staining Glass Dish Set with Stainless Steel Rack, 105 x 95 x 100 mm
SDP
Catalog No. NC1450892
Encompass_Preferred
A variety of inexpensive staining dishes made from molded glass with and without fitted glass and Stainless Steel lids. Ideal for immersing slides in several solutions. Holds 30 slides. Includes staining dish, glass cover, and slide rack.
Size: 105 x 95 x 100 mm
Catalog No. NC1450892 Supplier Electron Microscopy Sciences Supplier No. 71427
A variety of inexpensive staining dishes made from molded glass with and without fitted glass and Stainless Steel lids. Ideal for immersing slides in several solutions. Holds 30 slides. Includes staining dish, glass cover, and slide rack.
Size: 105 x 95 x 100 mm