Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Set of Flint Bottles with Dropper
GREENER_CHOICESCI_ED

Catalog No. S81681 Shop All Supertek Scientific Products
Change view
Click to view available options
Capacity (Metric):
30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S81681 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S81681 Supplier Supertek™ Supplier No. CH99900SP
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

High quality and durable, Boston flint bottles with droppers have countless classroom uses.

  • High quality and durable
  • Boston round amber flint bottles with droppers
  • Countless classroom uses
  • Glass is non-porous allowing for easy cleaning or autoclaving
  • Pack of 12

Specifications

Capacity (Metric) 30 mL
Color Amber
Material Borosilicate Glass
Quantity 12
Sterility Non-sterile
Closure Included Yes
Graduated No
Chemical Label Blank
Autoclavable Autoclavable
Capacity (English) 1 oz.
Shape Round
Disposable Reusable
Product Type Boston Flint Dropping Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.