Learn More
Air-Tite Products SET™Inject Luer Slip Syringe
Catalog No. 14817267 Shop All Air-Tite Products
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
50 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-817-267 5 mL
14-817-263 1 mL
14-817-265 2 mL
14-817-262 10 mL
14-817-264 20 mL
14-817-266 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
NO
Catalog No. 14-817-267 Supplier Air-Tite Products Supplier No. RS5SET
Add to Cart
Add to Cart
NO
SET™ inject 3-Part Syringes, manufactured in Turkey and 510K registered for human medical use, are designed for precision, safety, and comfort.
  • Made from polypropylene with glass-like transparency, they feature a latex-free isoprene rubber gasket and silicone oil lubrication for smooth piston movement, along with a stop-point mechanism to prevent accidental removal.
  • Their ergonomic design allows single-handed use, while high impermeability ensures accurate dosing.
  • Each syringe is sterilized with ethylene oxide and individually packed in medical-grade blister packs with transparent film.
Specifications
Autoclavable Not Autoclavable
Material Polypropylene (Barrel), Polyisoprene Rubber (Tip), Polyethylene
Needle Gauge No Needle Included
Latex Free Yes
Product Type Syringe
Disposable Single-use
Volume (Metric) 5 mL
Connection Type Centric Luer Lock
Graduated Yes
Sterility Sterile
Not Registered with Health Canada for Human Medical Use