Learn More
World Precision Instrument Set of 11 Calcium Buffers
SDP
Catalog No. 502521209
Encompass
Use with calcium fluorescent indicators
CALBUF-2 is especially suitable for calibrating fluorescent Ca++ indicators. It provides eleven buffer standards in the 10-4 to 10-8 M Ca++ range, whereas other commonly used fluorescent Ca++ indicators have the apparent Kd in the range of 100 to 300 nM. As with any ionic sensitive indicator, the sensitivity range of these indicators is about 1.0 log unit above and below the Kd. CALBUF-2 provides seven calibration points in this sensitivity range. It has an osmolarity of 0.305, which is isotonic with most mammalian cells.
Catalog No. 50-252-1209 Supplier World Precision Instrument Supplier No. CALBUF2
Use with calcium fluorescent indicators
CALBUF-2 is especially suitable for calibrating fluorescent Ca++ indicators. It provides eleven buffer standards in the 10-4 to 10-8 M Ca++ range, whereas other commonly used fluorescent Ca++ indicators have the apparent Kd in the range of 100 to 300 nM. As with any ionic sensitive indicator, the sensitivity range of these indicators is about 1.0 log unit above and below the Kd. CALBUF-2 provides seven calibration points in this sensitivity range. It has an osmolarity of 0.305, which is isotonic with most mammalian cells.