Learn More
Electron Microscopy Sciences Citifluor™ Mountant Solution CFPVOH + AF100, Glycerol Free, Non Hardening 30 ML
SDP
Catalog No. 5030239
Encompass_Preferred
Citifluor mounting medium for immunofluorescence is designed to reduce the fading of the fluorescence of dyes used for labeling biological specimens. In the technique of fluorescence microscopy, the fluorescence is stimulated by high intensity UV and/or visible light. Absorption of the light populates an excited state of the dye (usually a singlet state) and this leads to fluorescence. However, the excited state of the dye may undergo chemical reaction, which leads to destruction as evidenced by the fading of the fluorescence and consequent loss of the image. Aqueous solution of a carefully selected PVOH containing an amine-based antifadent.
Material: Glycerol Free
Type: Non Hardening
Catalog No. 50-302-39 Supplier Electron Microscopy Sciences Supplier No. 1797835
Citifluor mounting medium for immunofluorescence is designed to reduce the fading of the fluorescence of dyes used for labeling biological specimens. In the technique of fluorescence microscopy, the fluorescence is stimulated by high intensity UV and/or visible light. Absorption of the light populates an excited state of the dye (usually a singlet state) and this leads to fluorescence. However, the excited state of the dye may undergo chemical reaction, which leads to destruction as evidenced by the fading of the fluorescence and consequent loss of the image. Aqueous solution of a carefully selected PVOH containing an amine-based antifadent.
Material: Glycerol Free
Type: Non Hardening