Learn More
Perkin Elmer US LLC Set of UV/Vis Cell - 40mm
Catalog No. 502112611
Encompass_Preferred
This set of UV/Vis Cells includes two rectangular cells made of quartz. These cells have a pathlength of 40 mm.
Catalog No. 50-211-2611 Supplier Perkin Elmer US LLC Supplier No. N9302705
Add to Cart
Add to Cart
Request a Quote
This set of UV/Vis Cells includes two rectangular cells made of quartz. These cells have a pathlength of 40 mm.