Learn More
Thermo Scientific VF-P20 PTFE Pump Head Seals
Supplier:  Thermo Scientific 6040.2208
Encompass
PTFE seal engineered as leak-proof high pressure and temperature watertight pump seal with chemical resistance.
Catalog No. 50-134-7797