Learn More
Waters Corp Waters Setup Solution, QP, SQD, Mass Spectrometry
Catalog No. 50792837 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This setup solution is designed for use with mass spectrometry instruments. It facilitates the infusion and setup processes, specifically for entering masses during ESI positive and ESI negative modes.
  • Used for infusing and setting up mass spectrometry instruments
  • Compatible with instruments such as SQD, ACQUITY TQD, Quattro Premier, Quattro Premier XE, and Quattro micro
  • Provides specific mass points for ESI positive mode: 175.1, 455.3, 1080.8, 2034.6
  • Provides specific mass point for ESI negative mode: 554
Catalog No. 50-792-837 Supplier Waters Corp Supplier No. 700003105
This setup solution is designed for use with mass spectrometry instruments. It facilitates the infusion and setup processes, specifically for entering masses during ESI positive and ESI negative modes.
  • Used for infusing and setting up mass spectrometry instruments
  • Compatible with instruments such as SQD, ACQUITY TQD, Quattro Premier, Quattro Premier XE, and Quattro micro
  • Provides specific mass points for ESI positive mode: 175.1, 455.3, 1080.8, 2034.6
  • Provides specific mass point for ESI negative mode: 554