Learn More
Enzo Life Sciences SET7/9 (human), (recombinant) (100µg)
SDP
Catalog No. 502007945
Encompass_Preferred
Alternative name: Histone H3 K4 methyltransferase. Concentration: 1mg/ml. Endotoxin: <1.0EU/µg protein (LAL test). Purity: ≥95% (SDS-PAGE). MW: ~40.7kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 0.2M sodium chloride, 5mM DTT and 20% glycerol. Source: Produced in E. coli. Long Term Storage: -20°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7945 Supplier Enzo Life Sciences Supplier No. ALX201178C100
Alternative name: Histone H3 K4 methyltransferase. Concentration: 1mg/ml. Endotoxin: <1.0EU/µg protein (LAL test). Purity: ≥95% (SDS-PAGE). MW: ~40.7kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 0.2M sodium chloride, 5mM DTT and 20% glycerol. Source: Produced in E. coli. Long Term Storage: -20°C. Handling: Avoid freeze/thaw cycles.