Learn More
SETA BIOMEDICALS Extremely bright, water-soluble, squaraine rotaxane (SR) label
Catalog No. NC1726436
Encompass
Extremely bright, water-soluble, amine-reactive squaraine rotaxane (SR) label containing one NHS-ester group. The ideal label for proteins and other aminomodified biomolecules including oligonucleotides.
Catalog No. NC1726436 Supplier SETA BIOMEDICALS Supplier No. K94149
Extremely bright, water-soluble, amine-reactive squaraine rotaxane (SR) label containing one NHS-ester group. The ideal label for proteins and other aminomodified biomolecules including oligonucleotides.