Learn More
Thermo Scientific Capillary Kit MP35N VF-P20 LP for Thermo Sci 3P Parts & Accessories, Set of 1
Catalog No. 501347839 Shop All Thermo Scientific Products
Encompass
Capillary columns of nickel-cobalt alloy MP35N with 0.20mm internal diameter and 5um film thickness provide researchers with low-polarity columns suited for separation of volatile organic compounds to analyze samples by gas chromatography.
Catalog No. 50-134-7839 Supplier Thermo Scientific Supplier No. 6040.3003
Capillary columns of nickel-cobalt alloy MP35N with 0.20mm internal diameter and 5um film thickness provide researchers with low-polarity columns suited for separation of volatile organic compounds to analyze samples by gas chromatography.