Learn More
Epigentek Inc SETD2 Polyclonal Antibody (100 ul)
SDP
Supplier:  Epigentek Inc A3720100
Encompass_Preferred
Unconjugated Polyclonal Antibody Raised in Rabbit. Immunogen: Synthetic Peptide of Human MonoMethyl-Histone H3-K14. Tested Applications: WB, IHC, IF. Reactivity: Human, Mouse, Rat, Broad Range. Alternative Names: H3t, H3.4, H3/g, H3FT.
Catalog No. 50-109-5420